Learn More
Stemcell Technologies Inc Phalloidin, iFluor™ 488, Size: 300 Tests
Catalog No. 502231579
Encompass
Phalloidin is a heptapeptide derived from the poisonous mushroom Amanita phalloides. Phalloidin binds with a high specificity to F-actin filaments in cells, preventing filament depolymerization. Fluorescently labeled phalloidin can be used in labeling, identification, and quantitation of F-actin in formaldehyde-fixed and permeabilized cells. Fluorescently labeled phalloidin has lower non-specific binding compared to actin antibodies across most species of plants and animals, which provides low background and high contrast during imaging.
Catalog No. 502231579 Supplier Stemcell Technologies Inc Supplier No. 1000852
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Phalloidin is a heptapeptide derived from the poisonous mushroom Amanita phalloides. Phalloidin binds with a high specificity to F-actin filaments in cells, preventing filament depolymerization. Fluorescently labeled phalloidin can be used in labeling, identification, and quantitation of F-actin in formaldehyde-fixed and permeabilized cells. Fluorescently labeled phalloidin has lower non-specific binding compared to actin antibodies across most species of plants and animals, which provides low background and high contrast during imaging.