missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Fluorescent Brightener 28

Fluoresces clear bluish under UV radiation. Fluorescent Brightener 28 is a colorless organic compound that is used for the staining of fungi and as a viability stain. Industrial uses include use as a fluorescent brightening agent for cellulose and polyamide fabrics, paper and detergents and soaps.

$288.28 - $288.28

Specifications

Storage Requirements Store at Room Temperature (15°C to 30°C)
Format Powder
Type Fluorescent Brightener
Concentration 0.05% to 0.20%
For Use With (Application) Fluorescent Brightener 28 is used for the staining of fungi and as a viability stain. Industrial uses include use as a fluorescent brightening agent for cellulose and polyamide fabrics, paper and in detergents and soaps.
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MP215806705
View Documents
MP Biomedicals™
0215806705
5 g
Each for $288.28
Only null left
 
Description

Description

Fluoresces clear bluish under UV radiation. Fluorescent Brightener 28 is a colorless organic compound that is used for the staining of fungi and as a viability stain. Industrial uses include use as a fluorescent brightening agent for cellulose and polyamide fabrics, paper and detergents and soaps.

  • This product is related structurally to Fluorescent Brightner 9 (Calcofluor White MR), which was used by Jefferies and Belcher and by Maeda and Ischida in plant tissue studies.
Specifications

Specifications

Store at Room Temperature (15°C to 30°C)
Fluorescent Brightener
Fluorescent Brightener 28 is used for the staining of fungi and as a viability stain. Industrial uses include use as a fluorescent brightening agent for cellulose and polyamide fabrics, paper and in detergents and soaps.
290°C
C40H42N12O10S2Na2
Calcofluor White M2R, Tinopal UNPA-GX
Powder
0.05% to 0.20%
Yellow
4404-43-7
960.96
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.