missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Fluoresceinamine isomer I, pure

CAS: 3326-34-9 | C20H13NO5 | 347.33 g/mol

Chemical Identifiers

CAS 3326-34-9
Molecular Formula C20H13NO5
Molecular Weight (g/mol) 347.33
MDL Number MFCD00005052
InChI Key GZAJOEGTZDUSKS-UHFFFAOYSA-N
Synonym 5-Aminofluorescein
PubChem CID 76845
IUPAC Name 6-amino-3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one
SMILES NC1=CC=C2C(=C1)C(=O)OC21C2=CC=C(O)C=C2OC2=CC(O)=CC=C12
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC400770010
View Documents Promotion Details
Thermo Scientific Chemicals
400770010
1 g Glass bottle N/A
Only null left
 
AC400770050
View Documents Promotion Details
Thermo Scientific Chemicals
400770050
5 g Glass bottle N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3326-34-9
347.33
GZAJOEGTZDUSKS-UHFFFAOYSA-N
76845
NC1=CC=C2C(=C1)C(=O)OC21C2=CC=C(O)C=C2OC2=CC(O)=CC=C12
C20H13NO5
MFCD00005052
5-Aminofluorescein
6-amino-3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one

Specifications

3326-34-9
C20H13NO5
19, V,8, 713
GZAJOEGTZDUSKS-UHFFFAOYSA-N
Authentic
347.33
347.32
Pure
Crystalline Powder
99%
MFCD00005052
5-Aminofluorescein
NC1=CC=C2C(=C1)C(=O)OC21C2=CC=C(O)C=C2OC2=CC(O)=CC=C12
6-amino-3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one
76845
95% (typically)
Brown/Red
Fluoresceinamine isomer I, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 222-043-6

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only