missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluoresceinamine, isomer I, 96%
Description
5-aminofluorescein is also used to prepare FITC Isomer I (CDX-F0011, CDX-F0020). It is a chemical used as molecular probe and important as fluorescent dye and for derivatives. Fluorescent labeling reagent for proteins. Used in the fluorescent antibody technique for rapid identification of pathogens.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-aminofluorescein is also used to prepare FITC Isomer I (CDX-F0011, CDX-F0020). It is a chemical used as molecular probe and important as fluorescent dye and for derivatives. Fluorescent labeling reagent for proteins. Used in the fluorescent antibody technique for rapid identification of pathogens.
Solubility
Soluble in water (partly), methanol (10 mg/ml), and acetone.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Light and Moisture Sensitive
5-aminofluorescein is also used to prepare FITC Isomer I (CDX-F0011, CDX-F0020). It is a chemical used as molecular probe and important as fluorescent dye and for derivatives. Fluorescent labeling reagent for proteins. Used in the fluorescent antibody technique for rapid identification of pathogens.
Solubility
Soluble in water (partly), methanol (10 mg/ml), and acetone.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Light and Moisture Sensitive
Specifications
Specifications
| Melting Point | 223°C (decomposition) |
| Beilstein | 48395 |
| Color | Brown to Red |
| Solubility Information | Soluble in water (partly),methanol (10mg/ml),and acetone. |
| Quantity | 5 g |
| Physical Form | Powder |
| Formula Weight | 347.32 |
| Percent Purity | 96% |
| Chemical Name or Material | Fluoresceinamine, isomer I |
Safety and Handling
| Hazard Category | H315-H319-H335-H500 |
| Hazard Statement | GHS H Statement H302-H315-H319-H335 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 222-043-6 |
| TSCA | Yes |
| Recommended Storage | Keep cold |
RUO – Research Use Only