missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluorescein, Water Soluble, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Grade | Reagent |
| Packaging | Plastic Bottle |
| Color | Orange |
| Quantity | 500 g |
| Chemical Name or Material | Fluorescein |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.