missing translation for 'onlineSavingsMsg'
Learn More

Fluorescein, TCI America™

Catalog No. F009525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F009525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F009525G Supplier TCI America Supplier No. F009525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2321-07-5
Molecular Formula C20H12O5
Molecular Weight (g/mol) 332.31
MDL Number MFCD00005050
InChI Key GNBHRKFJIUUOQI-UHFFFAOYSA-N
Synonym Acid Yellow 73
PubChem CID 16850
ChEBI CHEBI:31624
IUPAC Name 3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one
SMILES C1=CC=C2C(=C1)C(=O)OC23C4=C(C=C(C=C4)O)OC5=C3C=CC(=C5)O

Specifications

Melting Point 315°C
Quantity 25 g
Physical Form Crystal/Powder
Formula Weight 332.31
Chemical Name or Material Fluorescein
EINECSNumber (5)-0671&(5)-1416&(5)-3060
RTECSNumber LM5075000
TSCA Yes