missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluorescein Sodium Salt, indicator, 98.5 to 100.5%, Honeywell Fluka™
Indicator, 98.5-100.5%
$155.34 - $3,798.08
Chemical Identifiers
| CAS | 518-47-8 |
|---|---|
| Molecular Formula | C20H10Na2O5 |
| Molecular Weight (g/mol) | 376.28 |
| MDL Number | MFCD00167039 |
| InChI Key | NJDNXYGOVLYJHP-UHFFFAOYSA-L |
| IUPAC Name | disodium 2-(6-oxido-3-oxo-3H-xanthen-9-yl)benzoate |
| SMILES | [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC([O-])=CC=C12 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
60045781
|
Honeywell-Fluka
28803100G |
100 g | Glass Bottle |
Each for $155.34
|
|
||||
|
60045782
|
Honeywell-Fluka
288031KG |
1 kg | Glass Bottle |
Each for $1,093.90
|
|
||||
|
60045783
|
Honeywell-Fluka
288034X1KG |
4 x 1 kg | Glass Bottle |
Case of 4 Each for $3,798.08
|
|
||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 518-47-8 | |
| 376.28 | |
| NJDNXYGOVLYJHP-UHFFFAOYSA-L | |
| [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC([O-])=CC=C12 |
| C20H10Na2O5 | |
| MFCD00167039 | |
| disodium 2-(6-oxido-3-oxo-3H-xanthen-9-yl)benzoate |
Specifications
| >300°C | |
| C20H10Na2O5 | |
| NONH for all modes of transport | |
| NJDNXYGOVLYJHP-UHFFFAOYSA-L | |
| disodium 2-(6-oxido-3-oxo-3H-xanthen-9-yl)benzoate | |
| 376.27g/mol | |
| Indicator | |
| Powder |
| 518-47-8 | |
| MFCD00167039 | |
| 3833041 | |
| [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC([O-])=CC=C12 | |
| 376.28 | |
| 98.5 to 100.5% | |
| Red | |
| Fluorescein Sodium Salt |
Safety and Handling
EINECSNumber : 208-253-0