missing translation for 'onlineSavingsMsg'
Learn More

Fluorescein Isothiocyanate (mixture of 5- and 6- isomers) 97.0+%, TCI America™

Catalog No. F07841G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07841G 1 g
F0784100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07841G Supplier TCI America Supplier No. F07841G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 27072-45-3
Molecular Formula C21H11NO5S
MDL Number MFCD00005063
Synonym Isothiocyanatofluorescein

Specifications

Quantity 1 g
Formula Weight 389.38
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Fluorescein Isothiocyanate (mixture of 5- and 6- isomers)
TSCA Yes
Recommended Storage Refrigerator