Learn More
Thermo Scientific Chemicals Fluorescein isothiocyanate, isomer 1, 95%
CAS: 3326-32-7 | C21H11NO5S | 389.381 g/mol
Supplier: Thermo Scientific Chemicals L09319MD
Description
Fluorescein isothiocyanate is used as fluorescent labeling reagent for proteins and for rapid identification of pathogens in fluorescent antibody technique. It is also used as a reagent for microsequencing of proteins and peptides (HPLC). On celite it is used for the labeling of fibrinogen.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsFluorescein isothiocyanate is used as fluorescent labeling reagent for proteins and for rapid identification of pathogens in fluorescent antibody technique. It is also used as a reagent for microsequencing of proteins and peptides (HPLC). On celite it is used for the labeling of fibrinogen.
Solubility
Slightly soluble in DMSO, ethanol, methanol DMF and acetone
Notes
Incompatible with strong oxidizing agents, strong bases and amines.
Specifications
| Fluorescein isothiocyanate | |
| 3326-32-7 | |
| C21H11NO5S | |
| 250 mg | |
| Moisture sensitive | |
| Slightly soluble in DMSO,ethanol,methanol DMF and acetone | |
| C1=CC2=C(C=C1N=C=S)C(=O)OC23C4=C(C=C(C=C4)O)OC5=C3C=CC(=C5)O | |
| 389.381 | |
| CHEBI:37918 | |
| 95% |
| isomer 1 | |
| >360°C | |
| MFCD00005063 | |
| 1407295 | |
| fitc, fluorescein isothiocyanate isomer i, 5-isothiocyanatofluorescein, fluorescein 5-isothiocyanate, 5-fitc, fluorescein-5-isothiocyanate, fluorescein 5 6-isothiocyanate, fitc isomer 1, unii-i223nx31w9, fitc isomer | |
| MHMNJMPURVTYEJ-UHFFFAOYSA-N | |
| 3',6'-dihydroxy-6-isothiocyanatospiro[2-benzofuran-3,9'-xanthene]-1-one | |
| 18730 | |
| 389.39 |
Chemical Identifiers
| 3326-32-7 | |
| 389.381 | |
| MHMNJMPURVTYEJ-UHFFFAOYSA-N | |
| 18730 | |
| 3',6'-dihydroxy-6-isothiocyanatospiro[2-benzofuran-3,9'-xanthene]-1-one |
| C21H11NO5S | |
| MFCD00005063 | |
| fitc, fluorescein isothiocyanate isomer i, 5-isothiocyanatofluorescein, fluorescein 5-isothiocyanate, 5-fitc, fluorescein-5-isothiocyanate, fluorescein 5 6-isothiocyanate, fitc isomer 1, unii-i223nx31w9, fitc isomer | |
| CHEBI:37918 | |
| C1=CC2=C(C=C1N=C=S)C(=O)OC23C4=C(C=C(C=C4)O)OC5=C3C=CC(=C5)O |
Safety and Handling
GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P330-P333+P313-P342+P311-P362-P501c
H302-H315-H317-H319-H334-H335
EINECSNumber : 222-042-0
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only