missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Fluorescein isothiocyanate, isomer 1, 95%

Catalog No. AAL09319MF
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL09319MF 50 mg
AAL09319MD 250 mg
AAL0931903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL09319MF Supplier Thermo Scientific Chemicals Supplier No. L09319MF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3326-32-7 | C21H11NO5S | 389.381 g/mol

Fluorescein isothiocyanate is used as fluorescent labeling reagent for proteins and for rapid identification of pathogens in fluorescent antibody technique. It is also used as a reagent for microsequencing of proteins and peptides (HPLC). On celite it is used for the labeling of fibrinogen.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fluorescein isothiocyanate is used as fluorescent labeling reagent for proteins and for rapid identification of pathogens in fluorescent antibody technique. It is also used as a reagent for microsequencing of proteins and peptides (HPLC). On celite it is used for the labeling of fibrinogen.

Solubility
Slightly soluble in DMSO, ethanol, methanol DMF and acetone

Notes
Incompatible with strong oxidizing agents, strong bases and amines.

Chemical Identifiers

CAS 3326-32-7
Molecular Formula C21H11NO5S
Molecular Weight (g/mol) 389.381
MDL Number MFCD00005063
InChI Key MHMNJMPURVTYEJ-UHFFFAOYSA-N
Synonym fitc, fluorescein isothiocyanate isomer i, 5-isothiocyanatofluorescein, fluorescein 5-isothiocyanate, 5-fitc, fluorescein-5-isothiocyanate, fluorescein 5 6-isothiocyanate, fitc isomer 1, unii-i223nx31w9, fitc isomer
PubChem CID 18730
ChEBI CHEBI:37918
IUPAC Name 3',6'-dihydroxy-6-isothiocyanatospiro[2-benzofuran-3,9'-xanthene]-1-one
SMILES C1=CC2=C(C=C1N=C=S)C(=O)OC23C4=C(C=C(C=C4)O)OC5=C3C=CC(=C5)O

Specifications

Melting Point >360°C
Quantity 50 mg
Beilstein 1407295
Sensitivity Moisture sensitive
Solubility Information Slightly soluble in DMSO,ethanol,methanol DMF and acetone
Formula Weight 389.39
Percent Purity 95%
Chemical Name or Material Fluorescein isothiocyanate, isomer 1
Hazard Category H302-H315-H317-H319-H334-H335
Hazard Statement GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P330-P333+P313-P342+P311-P362-P501c
EINECSNumber 222-042-0
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only