missing translation for 'onlineSavingsMsg'
Learn More

Fluorescéine 5-isothiocyanate (isomère I) 97,0+%, TCI America™

Numéro de catalogue. p-7129447
Modifier l'affichage
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
F0026100MG 100 mg
F00261G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. F0026100MG Fournisseur TCI America Code fournisseur F0026100MG

Identifiants chimiques

CAS 3326-32-7
Molecular Formula C21H11NO5S
Molecular Weight (g/mol) 389.381
InChI Key MHMNJMPURVTYEJ-UHFFFAOYSA-N
Synonym Fluorescein Isothiocyanate, 5-Isothiocyanatofluorescein, 5-FITC
PubChem CID 18730
ChEBI CHEBI:37918
IUPAC Name 3',6'-dihydroxy-6-isothiocyanatospiro[2-benzofuran-3,9'-xanthène]-1-un
SMILES C1=CC2=C(C=C1N=C=S)C(=O)OC23C4=C(C=C(C=C4)O)OC5=C3C=CC(=C5)O

Spécifications

Formula Weight 389.38
Percent Purity ≥97.0% (HPLC,T)
Quantity 100 mg
Physical Form Cristal/Poudre
Chemical Name or Material Fluorescein 5-Isothiocyanate (isomer I)
TSCA Yes
Recommended Storage Réfrigérateur