missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Fluorescein, pure

Catalog No. p-133544
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC119240250 25 g Glass bottle
AC119241000 100 g Glass bottle
AC119245000 500 g Glass bottle
AC119240025 2.5 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC119240250 Supplier Thermo Scientific Chemicals Supplier No. 119240250
Only null left

CAS: 2321-07-5 | C20H12O5 | 332.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2321-07-5
Molecular Formula C20H12O5
Molecular Weight (g/mol) 332.31
MDL Number MFCD00005050
InChI Key GNBHRKFJIUUOQI-UHFFFAOYSA-N
PubChem CID 16850
ChEBI CHEBI:31624
IUPAC Name 3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one
SMILES C1=CC=C2C(=C1)C(=O)OC23C4=C(C=C(C=C4)O)OC5=C3C=CC(=C5)O

Specifications

Melting Point 314.0°C to 316.0°C
Beilstein 19, V,6, 456
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in dilute alkalies and dilute acids, soluble in boiling alcohol and ether, insoluble in benzene and chloroform
Formula Weight 332.3
Grade Pure
Packaging Glass bottle
Quantity 25 g
Chemical Name or Material Fluorescein
Hazard Category Warning
Hazard Statement GHS H Statement:
May cause an allergic skin reaction.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear eye protection/face protection.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 219-031-8
RTECSNumber LM5075000
TSCA TSCA

RUO – Research Use Only