missing translation for 'onlineSavingsMsg'
Learn More

Fluorescamine [for HPLC Labeling], TCI America™

Catalog No. p-7129530
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0192100MG 100 mg
F01921G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F0192100MG Supplier TCI America Supplier No. F0192100MG
Only null left

Chemical Identifiers

Molecular Formula C17H10O4
MDL Number MFCD00005928

Specifications

CAS 38183-12-9
Melting Point 155.0°C to 159.0°C
Quantity 100 mg
Synonym 4-Phenylspiro[furan-2(3H),1′-phthalan]-3,3′-dione
Formula Weight 278.26
Physical Form Crystalline Powder