missing translation for 'onlineSavingsMsg'
Learn More

Fluorene 95.0+%, TCI America™

Catalog No. p-7129443
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F001725G 25 g
F0017500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F001725G Supplier TCI America Supplier No. F001725G
Only null left

Chemical Identifiers

CAS 86-73-7
Molecular Formula C13H10
Molecular Weight (g/mol) 166.22
MDL Number MFCD00001111
InChI Key NIHNNTQXNPWCJQ-UHFFFAOYSA-N
Synonym fluorene, diphenylenemethane, o-biphenylenemethane, 2,3-benzindene, 2,2'-methylenebiphenyl, o-biphenylmethane, methane, diphenylene, fluoren, fluorenyl radical, flourene
PubChem CID 6853
ChEBI CHEBI:28266
IUPAC Name 9H-fluorene
SMILES C1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Physical Form Crystal/Powder at 20°C
Boiling Point 295°C
Melting Point 115°C
Quantity 25 g
Color White
UN Number 3077
Formula Weight 166.22
Percent Purity ≥95.0% (GC)
Chemical Name or Material Fluorene
EINECSNumber (4)-0643
RTECSNumber LL5670000
TSCA Yes