missing translation for 'onlineSavingsMsg'
Learn More

Fluoral-P (=4-Amino-3-penten-2-one) 98.0+%, TCI America™

Catalog No. p-7118272
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A53501G 1 g
A53505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A53501G Supplier TCI America Supplier No. A53501G
Only null left

Chemical Identifiers

CAS 1118-66-7
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.133
MDL Number MFCD00043715
InChI Key OSLAYKKXCYSJSF-ONEGZZNKSA-N
Synonym acetylacetonamine, fluoral-p, 4-amino-3-penten-2-one, e-4-aminopent-3-en-2-one, 3-penten-2-one, 4-amino, 3e-4-aminopent-3-en-2-one, 4-aminopent-3-en-2-one, 4-amino-pent-3-en-2-one, e-4-amino-3-pentene-2-one, 2-acetyl-1-amino-1-methylethylene
PubChem CID 5367854
ChEBI CHEBI:51695
IUPAC Name (E)-4-aminopent-3-en-2-one
SMILES CC(=CC(=O)C)N

Specifications

Melting Point 41°C
Color White-Yellow
Boiling Point 214°C
Quantity 1 g
Formula Weight 99.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Fluoral-P (=4-Amino-3-penten-2-one) [Fluorimetric reagent for aldehydes.]
TSCA Yes
Recommended Storage Freezer