Learn More
Thermo Scientific™ Fluocinonide
Chemical Identifiers
| CAS | 356-12-7 |
|---|---|
| Molecular Formula | C26H32F2O7 |
| Molecular Weight (g/mol) | 494.53 |
| InChI Key | WJOHZNCJWYWUJD-IUGZLZTKSA-N |
| IUPAC Name | 2-[(1S,2S,4R,8S,9S,11S,12R,13S,19S)-12,19-difluoro-11-hydroxy-6,6,9,13-tetramethyl-16-oxo-5,7-dioxapentacyclo[10.8.0.0²,â¹.0â´,â¸.0¹³,¹â¸]icosa-14,17-dien-8-yl]-2-oxoethyl acetate |
| SMILES | CC(=O)OCC(=O)[C@@]12OC(C)(C)O[C@@H]1C[C@H]1[C@@H]3C[C@H](F)C4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]21C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC465450010
|
Thermo Scientific™
465450010 |
1 g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC465452500
|
Thermo Scientific™
465452500 |
250 mg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 356-12-7 | |
| 494.53 | |
| 2-[(1S,2S,4R,8S,9S,11S,12R,13S,19S)-12,19-difluoro-11-hydroxy-6,6,9,13-tetramethyl-16-oxo-5,7-dioxapentacyclo[10.8.0.0²,â¹.0â´,â¸.0¹³,¹â¸]icosa-14,17-dien-8-yl]-2-oxoethyl acetate |
| C26H32F2O7 | |
| WJOHZNCJWYWUJD-IUGZLZTKSA-N | |
| CC(=O)OCC(=O)[C@@]12OC(C)(C)O[C@@H]1C[C@H]1[C@@H]3C[C@H](F)C4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]21C |
Specifications
| 356-12-7 | |
| 1 g | |
| CC(=O)OCC(=O)[C@@]12OC(C)(C)O[C@@H]1C[C@H]1[C@@H]3C[C@H](F)C4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]21C | |
| 494.53 | |
| Fluocinonide |
| C26H32F2O7 | |
| WJOHZNCJWYWUJD-IUGZLZTKSA-N | |
| 2-[(1S,2S,4R,8S,9S,11S,12R,13S,19S)-12,19-difluoro-11-hydroxy-6,6,9,13-tetramethyl-16-oxo-5,7-dioxapentacyclo[10.8.0.0²,â¹.0â´,â¸.0¹³,¹â¸]icosa-14,17-dien-8-yl]-2-oxoethyl acetate | |
| 494.53 |
Safety and Handling
GHS H Statement Fatal if swallowed.
Suspected of damaging fertility or the unborn child if inhaled.
GHS P Statement Wash face, hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Rinse mouth.
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.
GHS Signal Word: Danger
EINECSNumber : 206-597-6
RUO â Research Use Only