missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Fluocinonide

Catalog No. AC465452500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC465452500 250 mg
AC465450010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC465452500 Supplier Thermo Scientific Chemicals Supplier No. 465452500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 356-12-7
Molecular Formula C26H32F2O7
Molecular Weight (g/mol) 494.53
InChI Key WJOHZNCJWYWUJD-IUGZLZTKSA-N
IUPAC Name 2-[(1S,2S,4R,8S,9S,11S,12R,13S,19S)-12,19-difluoro-11-hydroxy-6,6,9,13-tetramethyl-16-oxo-5,7-dioxapentacyclo[10.8.0.0²,⁹.0⁴,⁸.0¹³,¹⁸]icosa-14,17-dien-8-yl]-2-oxoethyl acetate
SMILES CC(=O)OCC(=O)[C@@]12OC(C)(C)O[C@@H]1C[C@H]1[C@@H]3C[C@H](F)C4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]21C

Specifications

Melting Point 296.0°C
Quantity 250 mg
Formula Weight 494.53
Chemical Name or Material Fluocinonide
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Fatal if swallowed.

Suspected of damaging fertility or the unborn child if inhaled.

Precautionary Statement GHS P Statement Wash face, hands and any exposed skin thoroughly after handling.

IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

Rinse mouth.

Obtain special instructions before use.

Wear protective gloves/protective clothing/eye protection/face protection.

IF exposed or concerned: Get medical advice/attention.

EINECSNumber 206-597-6

RUO – Research Use Only