missing translation for 'onlineSavingsMsg'
Learn More

Fluoboric Acid, Reagent, 48-50%, Spectrum™ Chemical

Catalog No. 18602763
Change view
Click to view available options
Quantity:
500 mL
4 L
6 x 500 mL
20 L
4 x 4 L
200 L
Packaging:
Drum
Poly Bottle
Poly Drum
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602763 200 L Drum
18602758 500 mL Poly Bottle
18602759 6 x 500 mL Poly Bottle
18602760 4 L Poly Bottle
18602761 4 x 4 L Poly Bottle
18602762 20 L Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18602763 Supplier Spectrum Chemical Mfg Cor Supplier No. F1075200LTBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

F1075, 16872-11-0, HBF4

Spectrum™ Chemical Fluoboric Acid, Reagent , is an inorganic compound that is named oxonium tetrafluoroboranuide, is classified as a strong acid that is corrosive and is used in organic chemistry as a catalyst as well as in metal plating where it is combined with organic reagents and serves as an electrolyte in the cathode treatment of the plating process. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.

Chemical Identifiers

CAS 16872-11-0
Molecular Formula BF4H
Molecular Weight (g/mol) 87.81
InChI Key ODGCEQLVLXJUCC-UHFFFAOYSA-O
IUPAC Name hydrogen tetrafluoroboranuide
SMILES [H+].F[B-](F)(F)F

Specifications

CAS Max % 100%
Packaging Drum
Quantity 200 L
UN Number UN1775
Percent Purity 48 to 50%
Grade Reagent