missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ FLUO 3/AM, Calbiochem™,

Catalog No. 3432421MG
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3432421MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 3432421MG Supplier MilliporeSigma™ Supplier No. 3432421MG

Membrane-permeable version of FLUO 3

Chemical Identifiers

CAS 121714-22-5
Molecular Formula C51H50Cl2N2O23
Synonym 1-[2-Amino-5-(2,7-dichloro-6-hydroxy-3-oxo-3H-xanthen-9-yl)]-2 Acid Pentaacetoxymethyl Ester

Specifications

Quantity 1 mg
Solubility Information Soluble in DSMO
Formula Weight 1129.9
Percent Purity ≥95%
Grade HPLC
Chemical Name or Material FLUO 3/AM
Recommended Storage Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above -20°C (-4°F).