missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell™ Fluka™ Non-Polar Silica Based SPE Columns
End-capped, Medical-grade polypropylene with PE frits
Supplier: Honeywell™ 9004
Description
- All columns are reversed phase and should be preconditioned with a polar solvent with the elution solvent being non-polar
- All columns are endcapped and have a surface area of 500m2/g
Chemical Identifiers
| 40 to 63 μm | |
| Yes | |
| 60 Å | |
| 500 m2/g |
Specifications
| 3 mL | |
| 40 to 63 μm | |
| Yes | |
| 50 |
| 500 mg | |
| Octadecyl (C18) | |
| 60 Å | |
| 500 m2/g |