missing translation for 'onlineSavingsMsg'
Learn More

Solstice Non-Polar Silica Based SPE Columns

Catalog No. 6002723
Change view
Click to view available options
Bed Weight:
100 mg
200 mg
500 mg
1000 mg
2000 mg
Column Capacity (Metric):
1 mL
3 mL
6 mL
Sorbent:
Ethyl (C2)
Octadecyl (C18)
Octyl (C8)
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bed Weight Column Capacity (Metric) Sorbent
6002723 100 mg 1 mL Octadecyl (C18)
6002724 100 mg 1 mL Octyl (C8)
6002737 200 mg 3 mL Ethyl (C2)
6002741 200 mg 3 mL Octadecyl (C18)
6002742 200 mg 3 mL Octyl (C8)
6002876 500 mg 3 mL Octadecyl (C18)
6002714 1000 mg 6 mL Octadecyl (C18)
6002715 1000 mg 6 mL Octyl (C8)
6002734 2000 mg 6 mL Octadecyl (C18)
6002872 500 mg 6 mL Ethyl (C2)
6002875 500 mg 6 mL Octadecyl (C18)
6002878 500 mg 6 mL Octyl (C8)
6002879 500 mg 3 mL Octyl (C8)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 6002723 Supplier Solstice Supplier No. HWL9000

End-capped, Medical-grade polypropylene with PE frits

  • All columns are reversed phase and should be preconditioned with a polar solvent with the elution solvent being non-polar
  • All columns are endcapped and have a surface area of 500m2/g

Specifications

Column Capacity (Metric) 1 mL
Bed Weight 100 mg
Particle Size 40 to 63 μm
Sorbent Octadecyl (C18)
Endcapped Yes
Pore Size 60 Å
Quantity 100
Surface Area (Metric) 500 m2/g