missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ FluidEase™ Pro ClipTip™ Electronic Pipette Charging Stand and Extension Kit

Catalog No. 01671125
Change view
Click to view available options
For Use With:
FluidEase Pipette Charging Stand
FluidEase Pipettes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With
01671125 FluidEase Pipettes
01671124 FluidEase Pipette Charging Stand
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01671125 Supplier Thermo Scientific™ Supplier No. 9420520
Only null left

The Thermo Scientific™ FluidEase™ Pro ClipTip™ electronic pipette charging stand enables simultaneous wireless charging of up to four pipettes and provides dedicated resting positions for short workflow pauses, keeping electronic pipettes powered, accessible, and organized in shared laboratories.

The Thermo Scientific™ FluidEase™ Pro ClipTip™ electronic pipette charging stand enables simultaneous wireless charging of up to four electronic pipettes without individual cables. Dedicated resting positions allow users to temporarily place pipettes with tips attached and liquid inside during short workflow interruptions. A modular extension design supports scalable charging capacity for shared laboratory environments.

Features:

  • Electronic Pipette Charging Stand for Shared Laboratories: Designed specifically for Thermo Scientific™ FluidEase™ Pro ClipTip™ electronic pipettes used in single- and multichannel workflows.
  • Simultaneous Charging of Up to Four Pipettes: Supports wireless charging of up to four electronic pipettes at the same time, reducing cable clutter and bench complexity.
  • Dedicated Charging and Resting Positions: Includes active charging positions and separate temporary resting positions for safely placing pipettes during short workflow pauses.
  • Temporary Resting with Tips and Liquid Inside: Allows users to briefly rest pipettes with tips attached and liquid retained when reaching for tubes, reagents, or plates.
  • Modular and Scalable Design: Extension kits enable charging capacity to scale from one to four pipettes as laboratory needs grow.
  • Organized Bench Footprint: Keeps electronic pipettes visible, accessible, and securely positioned to support efficient shared-lab operation.

Specifications

Color White
For Use With FluidEase Pipettes
Type Charging Stand
Includes FluidEase Charging stand base (1-position)