Learn More
Thermo Scientific™ Fluid Transfer Assembly, Y Set
Designed to aid many common shop floor processes such as sampling, transfer, and filling, a fit-for-purpose solution in the required tubing size and length can be easily selected from these designs.
$46.31 - $113.33
Specifications
| Tubing Style | 3 x TPE 374 |
|---|---|
| Product Type | Y Set |
| Tubing Size | 2 x 20 in. to 20 in. (Length) |
| Catalog Number | Mfr. No. | Tubing Style | Connector Types | Dimensions | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Tubing Style | Connector Types | Dimensions | Price | Quantity | |||
|
SH3110801
|
Thermo Scientific™
SH3110801 |
3 x TPE 374 | MPX body-insert | 1/4 in. ID x 7/16 in. wall |
Each for $67.34
|
|
|||
|
SH3110901
|
Thermo Scientific™
SH3110901 |
3 x TPE 374 | MPX insert-body | 1/4 in. ID x 7/16 in. wall |
Each for $68.13
|
|
|||
|
SH3110701
|
Thermo Scientific™
SH3110701 |
3 x TPE 374 | MPX insert-body | 3/8 in. ID x 5/8 in. wall |
Each for $75.41
|
|
|||
|
SH3110401
|
Thermo Scientific™
SH3110401 |
3 x TPE 374 | MPX body-insert | 1/2 in. ID x 3/4 in. wall |
Each for $94.03
|
|
|||
|
SH3111001
|
Thermo Scientific™
SH3111001 |
3 x TPE 374 | Luer | 1/8 in. ID x 1/4 in. wall |
Each for $46.31
|
|
|||
|
SH3110501
|
Thermo Scientific™
SH3110501 |
3 x TPE 374 | MPX insert-body | 1/2 in. ID x 3/4 in. wall |
Each for $95.94
|
|
|||
|
SH3110601
|
Thermo Scientific™
SH3110601 |
3 x TPE 374 | MPX body-insert | 3/8 in. ID x 5/8 in. wall |
Each for $113.33
|
|
|||
Description
Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard Labtainer BPCs, Productainer BPCs, single-use bioreactors, mixers, and fermentors right on the shop floor to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process.- One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
- Multiple manufacturing sites ensure security in product supply
- Produced under cGMP conditions in ISO 7 clean areas to ensure a high level of quality
- Characterized using ISO, USP, EP, and ASTM test methods to validate performance
Specifications
| 3 x TPE 374 | |
| Y Set | |
| 2 x 20 in. to 20 in. (Length) |