Learn More
Thermo Scientific™ Fluid Transfer Assembly, Extension Set
Designed to aid many common shop floor processes such as sampling, transfer, and filling, a fit-for-purpose solution in the required tubing size and length can be easily selected from these designs.
Supplier: Thermo Scientific™ SH31119.03
Description
Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard Labtainer BPCs, Productainer BPCs, single-use bioreactors, mixers, and fermentors right on the shop floor to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process.- One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
- Multiple manufacturing sites ensure security in product supply
- Produced under cGMP conditions in ISO 7 clean areas to ensure a high level of quality
- Characterized using ISO, USP, EP, and ASTM test methods to validate performance
- Irradiation dose of 25-40 kGy provides minimum SAL of 10-6 for product contact surfaces
Chemical Identifiers
| Extension Set |
Specifications
| 1/8 in. ID x 1/4 in. OD | |
| Luer | |
| TPE 374, 5 m (16.4 ft) |
| 3.22 mm (1/8 in.) ID; 6.35 mm (1/4 in.) OD; TPE 374; 5 m (16.4 ft); Luer | |
| Extension Set |