Learn More
Thermo Scientific™ Fluid Transfer Assembly
GREENER_CHOICE
Catalog No. SS00042I
Change view
Click to view available options
Port Detail:
1.9 cm Sanitary/MPX Body
1.9 cm Sanitary/MPX Insert
3.8 cm Sanitary/MPX Body
3.8 cm Sanitary/MPX Insert
CPC Steam-Thru/ Kleenpak Insert
CPC Steam-Thru/ReadyMate DAC
Lynx ST/Kleenpak Insert Connections
Lynx ST/ReadyMate DAC Connections
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Port Detail
SS00042I CPC Steam-Thru/ReadyMate DAC
SS00043I CPC Steam-Thru/ Kleenpak Insert
SS00045I Lynx ST/Kleenpak Insert Connections
SS00062I 3.8 cm Sanitary/MPX Body
SS00044I Lynx ST/ReadyMate DAC Connections
SS00063I 1.9 cm Sanitary/MPX Body
SS00064I 3.8 cm Sanitary/MPX Insert
SS00065I 1.9 cm Sanitary/MPX Insert
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. SS00042I Supplier Thermo Scientific™ Supplier No. SS00042I
Specialty Fluid Transfer Assembly products integrate easily within your existing process and equipment to maximize productivity, reduce cross-contamination and keep your process running smoothly.
Specialized connectors and tubing in our ready-designed assemblies provide an off-the-shelf solution that addresses many fluid transfer needs while delivering efficiency and increasing process integrity.
  • End treatments facilitate simple interconnection with existing process and equipment
  • Multiple manufacturing sites ensure security in product supply
  • Produced under cGMP conditions in ISO 7 clean areas to ensure a high level of quality
Specifications
Tubing Style 91.4cm of 12.7cm ID Silicone Tubing with CPC Steam-Thru II and ReadyMate DAC Connections
Port Detail CPC Steam-Thru/ReadyMate DAC
Product Type Fluid Transfer Assembly