Learn More
Thermo Scientific™ Fluid Thioglycollate Medium with Indicator, Dextrose Medium
Reduce prep time while isolating aerobes, anaerobes and microaerophilic organisms with this liquid medium with formats available that meet international industry guidelines.
Supplier: Thermo Scientific™ RE112640
Description
Cultivate and isolate aerobes, anaerobes and microaerophilic organisms using Thermo Scientific™ Fluid Thioglycollate Medium with Indicator, Dextrose Medium.
- For sterility testing according to USP (United States Pharmacopeia)
Specifications
| Industrial Reference: AOAC, BAM, COMPF, USP. | |
| United States | |
| Microbiology Media |
| WMB Lug | |
| Fluid Thioglycollate Medium | |
| 10/Pk. |