Learn More
Thermo Scientific™ Fluid Thioglycollate Medium with Indicator, Dextrose Medium
Catalog No. R117800
Change view
Click to view available options
Quantity:
10/Pk.
20/Pk.
Format:
16mm x 125mm Tube, Hungate Cap
Bottle
SB
WMB Lug
Media Type:
Fluid Thioglycollate Medium
Manufacturing Origin:
United States
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format Media Type Manufacturing Origin
R117800 20/Pk. 16mm x 125mm Tube, Hungate Cap Fluid Thioglycollate Medium United States
R112640 10/Pk. WMB Lug Fluid Thioglycollate Medium United States
R112641 10/Pk. SB Fluid Thioglycollate Medium United States
R112646 10/Pk. Bottle Fluid Thioglycollate Medium United States
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. R117800 Supplier Thermo Scientific™ Supplier No. RE117800
Reduce prep time while isolating aerobes, anaerobes and microaerophilic organisms with this liquid medium with formats available that meet international industry guidelines.
Cultivate and isolate aerobes, anaerobes and microaerophilic organisms using Thermo Scientific™ Fluid Thioglycollate Medium with Indicator, Dextrose Medium.
  • For sterility testing according to USP (United States Pharmacopeia)
Specifications
Certifications/Compliance Industrial Reference: AOAC, BAM, COMPF, USP.
Format 16mm x 125mm Tube, Hungate Cap
Manufacturing Origin United States
Media Type Fluid Thioglycollate Medium
Product Type Microbiology Media
Quantity 20/Pk.