missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Fludrocortisone acetate

Catalog No. p-7139368
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC462090010 1 g
AC462090050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC462090010 Supplier Thermo Scientific Chemicals Supplier No. 462090010

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 514-36-3
Molecular Formula C23H31FO6
Molecular Weight (g/mol) 422.49
InChI Key SYWHXTATXSMDSB-GSLJADNHSA-N
PubChem CID 124040734

Specifications

Melting Point 224°C to 226°C
Quantity 1 g
Solubility Information Solubility in water: .. Other solubilities: soluble in chloroform,ethanol,methanol
Formula Weight 422.49
Chemical Name or Material Fludrocortisone acetate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Suspected of damaging fertility or the unborn child.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.
Obtain special instructions before us
EINECSNumber 208-180-4

RUO – Research Use Only