missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluconazole Impurity H Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 150194-52-8 |
| Purity Grade Notes | These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available. |
| Synonym | (2 RS)-1-Bromo-2-(2,4-difluorophenyl)-3-(1 H-1,2,4-triazol-1-yl)propan-2-ol (Fluconazole Impurity H) |
| Type | Certified Reference Material |
| Quantity | 30 mg |
| Grade | Certified Reference Material |