missing translation for 'onlineSavingsMsg'
Learn More

Flucloxacillin sodium

Catalog No. AAJ66615MC
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66615MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ66615MC Supplier Thermo Scientific Chemicals Supplier No. J66615MC
Only null left

Targets primarily the cell wall of susceptible gram-positive bacteria including Staphylococcus aureus. | CAS: 1847-24-1 | C19H16ClFN3NaO5S | 475.85 g/mol

Used in clinical in vitro tests known as antimicrobial susceptibility tests. It is used in treatment of pneumonia, bone infections, skin infections, wound infections and burns infections.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Flucloxacillin sodium is used to treat infections caused by susceptible Gram-positive bacteria. It is used in clinical in vitro tests known as antimicrobial susceptibility tests. It is used in treatment of pneumonia, bone infections, skin infections, wound infections and burns infections.

Solubility
Soluble in water

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.

Specifications

Type Flucloxacillin Sodium
Physical Form Powder
CAS 1847-24-1
Melting Point 176°C to 178°C
Packaging Brown Glass Bottle
Quantity 100 mg
Flash Point >110°C (230°F)
Sensitivity Light sensitive
Molecular Formula C19H16ClFN3NaO5S
MDL Number MFCD01682108
Boiling Point 677°C
Solubility Information Soluble in water
InChI Key OTEANHMVDHZOPB-SLINCCQESA-M
SMILES [Na+].CC1=C(C(=O)N[C@H]2[C@H]3SC(C)(C)[C@@H](N3C2=O)C([O-])=O)C(=NO1)C1=C(F)C=CC=C1Cl
IUPAC Name sodium (2S,5R,6R)-6-[3-(2-chloro-6-fluorophenyl)-5-methyl-1,2-oxazole-4-amido]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Molecular Weight (g/mol) 475.85
PubChem CID 101643208
ChEBI CHEBI:31615
Formula Weight 475.85
Chemical Name or Material Flucloxacillin sodium
Show More Show Less
Hazard Category H317-H334
Precautionary Statement P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
EINECSNumber 217-428-0
RTECSNumber XH8680000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only