Learn More
Chemglass Inc 350mL Filter Funnel, Buchner, Medium Frit
Supplier:  Chemglass Inc CG140223
Encompass
350mL Filter Funnel, Buchner, Medium Frit. Funnel has a sealed-in glass fritted disc of the listed porosity. Useful wherever filter paper or membranes cannot withstand the chemical attack of the filtrate. Constructed with heavy wall stems and the top of the funnel is beaded for additional strength. Not recommended for use where pressure would exceed 15 psi.
Catalog No. CG140223
Only null left