Learn More
Anton Paar Flow Cell with Filling Funnel for Abbemat 300/350/500/550
Catalog No. NC1818543
Encompass
For routine analysis of low viscosity liquids. The sample is poured into the funnel and completely replaces the previous one so that a quick and comfortable sample exchange is possible.Suitable for use with Anton Paar refractometersAbbemat 300 item NC1818534,Abbemat 350 item NC1818535,Abbemat 500 item NC1818537,Abbemat 550 item NC1818539
Catalog No. NC1818543 Supplier Anton Paar Supplier No. ANP 101396
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For routine analysis of low viscosity liquids. The sample is poured into the funnel and completely replaces the previous one so that a quick and comfortable sample exchange is possible.Suitable for use with Anton Paar refractometersAbbemat 300 item NC1818534,Abbemat 350 item NC1818535,Abbemat 500 item NC1818537,Abbemat 550 item NC1818539