missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Florisil™ for the determination of hydrocarbon acc. to ISO 9377-2

Catalog No. 111012983
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012983 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012983 Supplier MilliporeSigma™ Supelco™ Supplier No. 03286100GF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For the determination of hydrocarbon acc. to ISO 9377-2

Florisil™, adsorbent for the separation of lipids is a commercially-prepared magnesia silica gel. It has a coarse mesh size which helps in rapid flow rate. Florisil™ is effective in promoting [1,3]-sigmatropic shift of myceophenolic acid related allyl phenyl ethers. Florisil™ cartridges are used in atmospheric sampling to assess possible interference from air contaminants.

Specifications

Pore Size 60 Å
Grade Standard
Quantity 100 g
Surface Area (Metric) 289 m2/g
Product Line Florisil
Physical Form Powder
MDL Number MFCD00078249
CAS 1343-88-0

Florisil is a registered trademark of U.S. Silica Company