missing translation for 'onlineSavingsMsg'
Learn More

Florisil, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 1343-88-0
Molecular Formula MgO3Si
Molecular Weight (g/mol) 100.39
InChI Key ZADYMNAVLSWLEQ-UHFFFAOYSA-N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
MK6877500
View Documents
Avantor Macron
687708
500g Glass N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

1343-88-0
MgO3Si
100.39
ZADYMNAVLSWLEQ-UHFFFAOYSA-N

Specifications

1343-88-0
ZADYMNAVLSWLEQ-UHFFFAOYSA-N
Florisil, 60-100 Mesh
MgO3Si
100.39
Videos
SDS
Documents

Documents