missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Florisil™, 60-100 mesh, for column chromatography
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for column chromatography
Specifications
Specifications
| Color | White |
| Physical Form | Fine Powder |
| Quantity | 250 g |
| Fieser | 01,393; 09,223 |
| Merck Index | 10, 5509 |
| Packaging | Glass Bottle |
| Particle Size | Particle size analysis (typical values): 60 mesh: 2.0%, 70 mesh: 20.0%, 80 mesh: 41.0%, 100 mesh: 30.0%, 140 mesh: 6.9% |
| Formula Weight | 100.39 |
| Chemical Name or Material | Florisil™, 60-100 mesh, for column chromatography |
RUO – Research Use Only