Learn More
Thermo Scientific Chemicals Florisil™, 100-200 mesh, for column chromatography
Catalog No. p-70307
Change view
Click to view available options
Quantity:
250 g
500 g
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC205442500 250 g
AC205445000 500 g
AC205440050 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC205442500 Supplier Thermo Scientific Chemicals Supplier No. 205442500
Only null left
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Particle Size 100/120 Mesh
Packaging Glass Bottle
Quantity 250 g
Material Florisil
For Use With (Application) Column Chromatography
RUO – Research Use Only