missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Floor Prep Solution

Catalog No. 17985488
Change view
Click to view available options
Diameter (English):
6 in.
12 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (English)
17985488 6 in.
17985490 12 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17985488 Supplier Brady™ Supplier No. 178946

Eco-friendly, ready-to-use cleaner prepares floors for strong tape adhesion and long-lasting, professional floor markings.

  • Purpose-formulated to remove dirt, dust, and light residue, ensuring surfaces are properly prepared for tape application.
  • Maximizes floor tape adhesion and durability to deliver long-lasting, professional results.
  • Eco-friendly and safe, non-hazardous formula supports sustainable cleaning practices.
  • Versatile cleaner suitable for a wide range of floor types and facility environments.
  • Convenient jug size ideal for large cleaning tasks and frequent use.

Specifications

Product Type Cleaner
Color-coded No
Form Liquid
Scent Original
Sterility Non-sterile
Diameter (English) 6 in.
Height (English) 12 in.
Width (English) 6 in.