missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ Float and Spin Magnet Demonstrator

Catalog No. S15959
Change view
Click to view available options
Grade:
5+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S15959 5+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S15959 Supplier United Scientific™ Supplier No. FLSPMAG

Engaging visual demonstration of magnetic repulsion.

  • This attention-getting demonstration of magnetic repulsion uses a clear acrylic plate that keeps a spinning axle oriented above magnets built into the base of the apparatus.
  • With a simple twist of your fingers, you can start this levitating magnet spinning and kick start a classroom discussion.
  • The black and white coloring makes the demonstration easy to see and helps track the speed of the rotation.
  • Easy to see black and white pattern
  • Smooth rotation
  • No batteries required
  • The axle measures 5 in. long Dimensions: 2.5 x 5 x 3 in.

Specifications

For Use With (Application) Magnetic Demonstrations
Grade 5+
Product Type Flip Spin Magnet

Warning: Cancer and reproductive harm