Perkin Elmer Canada UV Quartz SUPRASIL Rectangular Flow-Through Cell, Semi-Micro/Ultra-Micro with In- and Outlet Tubes - 160 L, Qty. 1
Catalog No. 509042645
The UV Quartz SUPRASIL Flow-Through Cell has a light path of 10 mm, a volume of 390 L, an aperture of 11 x 3.5 mm, and outside dimensions of 35 mm (H) x 12.5 mm (W) x 12.5 mm (D). Qty. 1
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The UV Quartz SUPRASIL Flow-Through Cell has a light path of 10 mm, a volume of 390 L, an aperture of 11 x 3.5 mm, and outside dimensions of 35 mm (H) x 12.5 mm (W) x 12.5 mm (D). Qty. 1