missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ FLJ10154 Recombinant Protein Antigen

Catalog No. NBP258390PE
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP258390PE 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP258390PE Supplier Novus Biologicals™ Supplier No. NBP258390PEP

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human FLJ10154. Source: E.coli Amino Acid Sequence: RSRSTNTAVSRRERDRERASSPPDRIDIFGRTVSKRSSLDEKQKR The FLJ10154 Recombinant Protein Antigen is derived from E. coli. The FLJ10154 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.

Specifications

Gene ID (Entrez) 55082
Purification Method >80% by SDS-PAGE and Coomassie blue staining
Common Name FLJ10154 Recombinant Protein Antigen
Content And Storage Store at −20C. Avoid freeze-thaw cycles.
Formulation PBS and 1M Urea, pH 7.4.
For Use With (Application) Blocking/Neutralizing, Control
Gene Alias arginine and glutamate rich 1, arginine and glutamate-rich protein 1, DKFZp686O08106, FLJ10154
Gene Symbol ARGLU1
Label Type Unlabeled
Product Type Recombinant Protein Antigen
Quantity 100 μL
Regulatory Status RUO
Source E.Coli
Specific Reactivity This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-50337. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml
Show More Show Less

For Research Use Only