missing translation for 'onlineSavingsMsg'
Learn More

Fischer Technical Company™ Flip Style Two Valve Natural Rubber Safety Pipet Filler

Catalog No. S012385
Encompass
Change view
Click to view available options
Color:
Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
S012385 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Prop 65: Cancer and Reproductive Harm.
Catalog No. S012385 Supplier Fischer Technical Company™ Supplier No. FLIP003

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Prop 65: Cancer and Reproductive Harm.

Safety Pipet Filler is easy to use and clean.

This patented ball fits pipets up to 50 mL.

  • Press on the bulb to evacuate
  • Press on the up arrow to intake solution
  • Drain by pressing on the down arrow
  • Top valve can be removed and reinserted for easy cleaning of the inside

Specifications

Autoclavable Autoclavable
Color Blue
Disposable Reusable
For Use With Pipette
Grip Type Round
Material Natural Rubber
Model Flip
Number of Channels 1
Sterility Non-sterile
Type Pipette Filler
Volume (Metric) 50 mL
Show More Show Less