Learn More
Restek™ Flip Seal™ Dual Vespel™ Ring Inlet Seals
Reversible, two-sided design allows significantly more analyses than other seals
$197.32 - $1107.83
Specifications
| Diameter (Metric) Inner | 1.2 mm |
|---|---|
| For Use With (Application) | Ded Design Allows Significantly More Analyses Than Other Seals |
| Catalog Number | Mfr. No. | Type | Packaging Quantity | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Type | Packaging Quantity | Price | Quantity | ||||
06-701-309
|
Restek™
RK23414 |
Siltek-Deactivated | 10/Pk. |
Each for $1,107.83
|
|
||||
06-701-306
|
Restek™
RK23411 |
Gold-Plated | 2/Pk. |
Each for $197.32
|
|
||||
06-701-308
|
Restek™
RK23413 |
Gold-Plated | 10/Pk. |
Each for $1,087.87
|
|
||||
Description
- Highly inert gold or Siltek™deactivated seals reduce breakdown and adsorption of active compounds, maximizing component transfer to GC column
- Vespel™ ring embedded in top and bottom surface eliminates need for washer
- Very little torque required to make seal, reduces operator variability
Specifications
| 1.2 mm | |
| Ded Design Allows Significantly More Analyses Than Other Seals |