missing translation for 'onlineSavingsMsg'
Learn More

Wells Lamont™ FlexTech™ Y9219 Polyurethane-Coated Cut-Resistant Gloves

Catalog No. 17200412
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17200412 Small
17200413 Medium
17200414 Large
17200415 X-Large
17200416 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17200412 Supplier Wells Lamont™ Supplier No. Y9219S
Only null left

Ultra-lightweight liner with polyurethane coating on the palm delivers dexterity, tactility and comfort. FlexTech™ Y9219 Polyurethane-Coated Cut-Resistant Gloves provide ANSI Cut Level A9 cut resistance for exceptional protection.

  • 18-gauge ultra lightweight liner delivers dexterity, tactility and comfort
  • Constructed from a blend of tungsten, fiberglass, HPPE and polyester
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Compatible with touchscreen devices for easy use with tablets and phones
  • Machine washable for extended wear life
  • ANSI A9 Cut Resistance
  • ANSI Puncture Level 4

Specifications

ANSI Abrasion Rating ANSI 4
Coating Material Polyurethane
Color Gray
Cut Resistance Rating ANSI A9
Hand Specific
Material HPPE, Polyester, Tungsten, Fiberglass
Product Type Cut-resistant Gloves
Size Small
For Use With (Application) Assembly, metal handling, fabrication and stamping
Glove Cuff Style Knit
Model Y9219S
Puncture Rating ANSI/ISEA 105-16 Level 4
Series FlexTech™ Y9219
Yarn Gauge 18
Show More Show Less