missing translation for 'onlineSavingsMsg'
Learn More

Wells Lamont Industrial FlexTech ANSI A4 Cut Resistant Gloves

Catalog No. p-7228431
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17010051 Small
17010052 Medium
17010053 Large
17010054 X-Large
17010055 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17010051 Supplier Wells Lamont Industrial Supplier No. Y9258SA
Only null left

FlexTech ANSI A4 Cut Resistant Gloves with a foam nitrile palm and ESD properties to protect equipment and processes.

  • Anti-static properties that rapidly neutralize the electric discharge for the worker and ESD properties that protect product equipment and workpieces
  • The ultra-lightweight liner provides excellent dexterity as an 18 gauge thin glove with an ANSI Cut Level A4 protection to reduce hand injuries and increased abrasion resistance and durability for prolonged use
  • Foam nitrile palm for grip and touchscreen compatibility
  • Offers ANSI 4 puncture and abrasion resistance

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance EN 16350:2014, EN 1149-2:1997
Coated Area Palm
Applications Industrial
Coating Material Nitrile Foam
Color Gray
Cut Resistance Rating ANSI A4
Disposable Reusable
Exterior Finish Fully Textured
Material Nylon, HPPE, Fiberglass, Lycra, Stainless Steel
Product Type Cut Resistant Glove
Size Small
For Use With (Application) Metal Stamping, Electronics Assembly, Small Parts Handling and Assembly
Model Y9258
Resistant To Cut Resistant
Yarn Gauge 18
Show More Show Less