missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Interscience FlexiPump
Description
FlexiPump is a peristaltic pump with enhanced accuracy for the dispensing of culture media, agar and any type of diluent. Compact and ergonomic, it comes with a user-friendly interface.
- Dispensing mode: Continuous, Dose, Multi-dose
- Dose volume: from 50 μL to 99.9 L
- Flow rate: Up to 1.25 L/min.
Specifications
Specifications
| Control Type | Analog |
| Display Type | Digital |
| Enclosure Rating | IP31 |
| Flow Rate | 1.5 mL/min. to 1.25 L/min. |
| Motor Speed | 10 to 550 rpm |
| Reversible | No |
| Model | FlexiPump |
| Voltage | 100/240 V |