Learn More
Ampac™ Flexibles SealPAK Heavy-Duty Pouches
Ideal for applications requiring long-term storage, sharper objects, or solutions of formaldehyde, acids, alkalies, greases, oils and organic solvents
Supplier: Ampac™ 504B1M
Description
Ideal for applications requiring long-term storage, sharper objects, or solutions of formaldehyde, acids, alkalies, greases, oils and organic solvents
- Boilable and freezable pouches can sustain temperatures from -56 to 115°C (-70 to 240°F)
- 4.5 mils thick
- Transparent, airtight and leak-proof
- Can be sterilized with gamma, infrared and e-beam
- Sealed pouches can also be surface sterilized using VHP
Order Info
Also available in heat-sealable tubular roll stock that allows you to create custom-size pouches.
Specifications
| 32 oz. | |
| 1000/Cs. | |
| 12 x 8 in. (30.5 x 20.3cm) | |
| 30.5 cm | |
| 20.3 cm |
| Polyester | |
| 4.50 mil | |
| 12 in. | |
| 8 in. |