missing translation for 'onlineSavingsMsg'
Learn More

SOMNI Scientific™ Flexible Tubing

Catalog No. p-7221164
Change view
Click to view available options
Color:
Blue
Green
Orange
Red
Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
04777180 Blue
04777182 Red
04777183 Orange
04777184 Yellow
04777185 Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 04777180 Supplier SOMNI Scientific™ Supplier No. TB2004
Only null left

Provided custom cut per 12-inch increments to your specification. Opaque and firm yet flexible. Colored coded tubing for easy identification.

  • Opaque flexible tubing provided custom cut per 12 inch increments to your specification
  • Tubing is 0.25 in. ID
  • Firm yet flexible PVC
  • Colored coded tubing for easy identification

Specifications

Diameter (English) 0.25 in.
For Use With (Application) Research Animal Inhalant Anesthesia
Color Blue