Learn More
BRANDTECH™ BRAND™ Flexible Discharge Tube with recirculation valve for all models of Dispensette™ S Bottletop Dispensers.
Fits all models of Dispensette™ S, Dispensette™ S Organic, and Dispensette™ S Trace Analysis, manufactured in 2016 or later.
Supplier: BRANDTECH™ BRAND™ 708134
Description
- Easily fill racks of tubes with Dispensette™ S, Dispensette™ S Organic, or Dispensette™ S Trace Analysis dispensers
- 800 mm PTFE tubing is conveniently coiled
- Textured safety handle for a secure grip
- Features an integrated recirculation valve
- For best results, use a bottle stand with Trace Analysis dispensers and with small or lightweight bottles
- Not for use with hydrofluoric acid (HF)
- Available in two sizes: 708132- For dispensers with a nominal capacity of 10 mL or smaller; 708134 - For dispensers with a nominal capacity of 25 mL or larger
Chemical Identifiers
| PTFE | |
| 800 mm |
Specifications
| Nominal Volume 25, 50, 100 mL | |
| PTFE | |
| 25, 50, 100 mL | |
| 800 mm |