Learn More
BRANDTECH™ BRAND™ Flexible Discharge Tube with recirculation valve for all models of Dispensette™ S Bottletop Dispensers.
Fits all models of Dispensette™ S, Dispensette™ S Organic, and Dispensette™ S Trace Analysis, manufactured in 2016 or later.
Supplier:  BRANDTECH™ BRAND™ 708132
 View more versions of this product
Catalog No. 14380909
Only null left
Description
Description
  • Easily fill racks of tubes with Dispensette™ S, Dispensette™ S Organic, or Dispensette™ S Trace Analysis dispensers
  • 800 mm PTFE tubing is conveniently coiled
  • Textured safety handle for a secure grip
  • Features an integrated recirculation valve
  • For best results, use a bottle stand with Trace Analysis dispensers and with small or lightweight bottles
  • Not for use with hydrofluoric acid (HF)
  • Available in two sizes: 708132- For dispensers with a nominal capacity of 10 mL or smaller; 708134 - For dispensers with a nominal capacity of 25 mL or larger
Specifications
Chemical Identifiers
PTFE
800 mm
Specifications
Nominal Volume 1, 2, 5, 10 mL
PTFE
1, 2, 5, 10 mL
800 mm
Product Suggestions
Product Suggestions