missing translation for 'onlineSavingsMsg'
Learn More

Cardinal Health™ Flexal™ Nitrile Gloves

Catalog No. 19066125
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19066125 X-Small
19066126 Small
19066127 Medium
19066128 Large
19066129 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19066125 Supplier Cardinal Health™ Supplier No. 88TN01XS

Nitrile medical gloves provide tactile sensitivity and protect hands from potentially infectious materials.

  • Textured fingertips improve grip even when wet
  • Tested for use with various lab chemicals and chemotherapy drugs
  • Latex-free
  • Powder free
  • Ambidextrous

Specifications

Certifications/Compliance ASTM D6319, ASTM D6978
Chemotherapeutic Rated Yes
Applications Healthcare, Laboratory
AQL 1
Color Blue
Disposable Single-use
Exterior Finish Partially Textured
Finger Thickness (English) 3.7 mil
Finger Thickness (Metric) 0.09 mm
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size X-Small
Thickness (English) 3.7 mil
Thickness (Metric) 0.10 mm
FDA 510 (k) Yes
Model 88TN01XS
Series Flexal™
Show More Show Less